Learn More
Hamilton™ Swivel Union 1/4-28 Threads
Product Code. 11502475
Change view
Click to view available options
Material:
CTFE
Unit Size:
Each
Product Code. Material Unit Size
11502475 CTFE Each
1 options
Product Code. 11502475 Supplier Hamilton™ Supplier No. 35037
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Specification Sheet
  • Connection 1 – Male 1/4-28
  • Connection 2 – Male 1/4-28
  • Fitting Material – CTFE
  • Sales Quantity – 1 each
  • I.D. – 1.5 mm
Specifications
Type Male 1/4-28
Material CTFE