Learn More
MedChemExpress SYBR Green qPCR Master Mix (Low ROX)
Product Code. 16428946
Encompass
Change view
Click to view available options
Quantity:
100 Reactions
Unit Size:
100 reactions
Product Code. Quantity Unit Size
16428946 100 Reactions 100 reactions
1 options
Product Code. 16428946 Supplier MedChemExpress Supplier No. HYK0522
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MedChemExpress SYBR Green qPCR Master Mix (Low ROX) is provided as a simple-to-use, stabilized 2X formulation that includes all components for qPCR except sample DNA, primers and water, in which Low ROX Reference Dye is included.
Specifications
Content And Storage Stored at -20°C protecting from light, and is stable for up to 18 months. Avoid repetitive freeze-thaw cycles while using. For immediate use, components may be stored at 2°C to 8°C.
Product Type Kits
Quantity 100 Reactions
Shipping Condition Room temperature