missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Syn-PER™ Synaptic Protein Extraction Reagent

Product Code. 12341963 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12341963 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12341963 Supplier Thermo Scientific™ Supplier No. 87793
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate synaptosomes containing functional synaptic proteins from neuronal tissue and primary cultured neurons with this cell lysis reagent.

Thermo Scientific™ Syn-PER Synaptic Protein Extraction Reagent is a proprietary cell lysis reagent for efficient isolation of synaptosomes containing functional synaptic proteins from neuronal tissue and primary cultured neurons.

Syn-PER Reagent is used to prepare synaptosomes containing biologically active pre-and post-synaptic proteins. When used with fresh neuronal tissue or primary cultured neurons, synaptosomes prepared with the Syn-PER Reagent can be used to study synaptic transmission. The synaptosomal proteins contained in these extracts can also be used for downstream applications such as Western blotting, immunoprecipitation, enzymatic activity assays and protein-protein interaction studies. Syn-PER Synaptic Protein Extraction Reagent minimizes the degradation of phosphoproteins and is ideal for studies requiring the preservation of phosphoprotein integrity.

Highlights:

  • Efficient extraction – obtain up to 10μg of synaptic protein per milligram of neuronal tissue or 4μg synaptic protein per 35mm dish of primary cultured neurons (106 cells)

  • Gentle formulation – isolate viable synaptosomes, extract native synaptic proteins and preserve phosphoprotein integrity

  • Fast procedure – obtain synaptosomal suspension in less than one hour

  • Simple protocol – requires no ultracentrifugation steps

Recommended for:

Isolate functional synaptosomes to study neurotransmitter release; Extract pre- and post-synaptic proteins to identify changes in protein composition and function in synapses; Preserve and study labile or transient protein phosphorylation events associated with synapses

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Format Liquid
Product Type Reagent
Volume (Metric) 100 mL
Content And Storage Upon receipt store product at 4°C.
Product Line Syn-PER

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.