Learn More
Tocris Bioscience™ SynaptoRed C2
GREENER_CHOICE
Fluorescent dye; stains synaptic vesicles
Brand:  Tocris Bioscience™ 5118/1
Encompass
Product Code. 15756486
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
SynaptoRedTM C2 is a fluorescent dye; stains synaptic vesicles. Becomes fluorescent when incorporated into plasma membrane, used to follow synaptic activity at the synapse or neuromuscular junction. Excitation and emission spectra are ∽ 515 and 640 nm respectively.
Specifications
Specifications
>95%