missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ G:BOX Chemi XX9 Gel Documentation System

Product Code. 15843280 Shop All Syngene Products
Change view
Click to view available options
Pixel Size:
9 MP
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pixel Size Unit Size
15843280 9 MP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15843280 Supplier Syngene™ Supplier No. GBOXCHEMIXX9E
  • ONLINE EXCLUSIVE PRICE
    33505.00 €
    30637.82 € / Pack of 1
    Save 2867.18 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-04-2027
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.

G:BOX Chemi XX9 Gel Documentation System offers high performance imaging of gels and chemiluminescence blots with incredible resolution.

  • Versatile gel documentation system for blots and gels.
  • Capable of many imaging applications including DNA/Protein gels, chemiluminescence, stain free gels, coloured multiplex fluorescence blots, 2D gels and In Vivo applications (additional accessories may be needed).
  • Includes a Peltier cooled, high resolution 9 MP camera, capable of producing 16 bit images (with 65535 grey scales).
  • The camera chip is a leader in the field with a QE of 73% at 425 nm (peak 77%).
  • The system has an F0.95 lens with motorized iris and focus (with feedback) for short acquisition times. The stretch darkroom has a unique motorized lift to utilize zoom features, with autofocus and ensuring the maximum image resolution of the gel or blot is captured.
  • Comes standard with a seven position filter wheel, epi white light and an optional LED gantry which gives capability for fluorescence imaging, for red, green, blue and near-IR imaging (additional accessories will need to be purchased). The maximum viewing area available is 32.3 x 25.6 cm.
  • PC required (not included).

    GeneSys is an application driven software designed for rapid image capture of biological samples. It has a simple icon based user interface usable on Win 10, 8 and 7 platforms. GeneSys allows the user to take the guess work out of imaging, and produce quick and simple publication quality images. All files produced are a unique SGD format, where only raw data is analysed for quantification of samples. Files can also be saved as TIFF, JPEG, BMP formats. User protocols can be saved to review previous images captured, and easily replicate future imaged gels. All captured images can be simply edited with annotations, edit tools, image sharpen and exported to GeneTools.

    GeneTools software provides the user with rapid quantification of samples and goes from blots to analysed data in a few quick steps.
    Analysis sample types include molecular weight, colony counting, spot blot and band matching.
    All samples can be quantified, including multiplexed protein fluorescence analysis of up to 5 samples at once. Raw data files can be quickly quantified to produce true data for publication data. High quality reports can be exported in Word and Excel files for simple review.

    Both GeneSys and GeneTools software meet the requirements for 21 CFR Part 11 and GLP environments.

    The system comes with a 3 year warranty (return to factory), unlimited licences of GeneTools analysis software and twice annual free software updates.

    IQ/OQ is available upon request.

gel, western, fluorescence, documentation, chemiluminescence, protein

Spécification

Description Gel Documentation System
Voltage 230 V
Depth (English) 21.7 in.
Depth (Metric) 55 cm
Filter Type 7-position Filter Wheel
Height (English) 38.9 in.
Filter Area 20 x 20 cm or 25 x 30 cm depending on transilluminator selected
Pixel Size 9 MP
Width (English) 22.4 in.
Height (Metric) 99 cm
Operating System Windows 7/8/10
Resolution 9 MP up to 27 MP
Software Genesys, Genetools
Transilluminator Optional
Wavelength Dependant on transilluminator
Width (Metric) 57 cm
Model Gbox-Chemi-XX9
Printer Included No (optional)
Camera Type 9 MP
Size 57 x 99 x 55 cm
For Use With (Application) Gel Documentation
Product Line Syngene
Product Type Gel Imaging System
Quantity 1 Ea.
Display Type None
Frequency 50 Hz
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.