Learn More
Thermo Scientific Chemicals Syntide 2, 96%
For research use
Brand: Thermo Scientific Chemicals J62843.#0
Code : NEW
Additional Details : CAS Number : 108334-68-5 Weight : 0.00005kg
Description
Selective substrate for calmodulin-dependent protein kinase II
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Enzyme Assay Substrate | |
5mg | |
RUO | |
Syntide 2, 96% | |
Lyophilized |
Lyophilized powder | |
-30° to -10°C | |
Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys | |
Native | |
96% |
RUO – Research Use Only