missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Syntide 2, 96%

Product Code. 15485149
Change view
Click to view available options
Quantity:
1mg
5mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485149 1mg 1mg
15475149 5mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485149 Supplier Thermo Scientific Chemicals Supplier No. J62843.M^
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For research use

Selective substrate for calmodulin-dependent protein kinase II

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Assay Substrate
Formulation Lyophilized powder
Quantity 1mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys
Product Type Syntide 2, 96%
Recombinant Native
Form Lyophilized
Purity or Quality Grade 96%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.