missing translation for 'onlineSavingsMsg'
Learn More

Syringaldazine 99+%

Product Code. 16574194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16574194 5 g 5g
16564194 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16574194

Brand: Thermo Scientific Acros 175930050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14414-32-5 | C18H20N2O6 | 360.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14414-32-5
Molecular Formula C18H20N2O6
InChI Key MYNVJNZGTAVRJD-UHFFFAOYSA-N
Synonym 4-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, 3,5-dimethoxy-4-hydroxybenzalazine, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone, 4,4'-hydrazine-1,2-diylidenebis methanylylidene bis 2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 2-4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone, 4-hydroxy-3,5-dimethoxybenzaldehyde azine, syringaldazine, 4-1e-e-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol
PubChem CID 5379425

Specifications

Melting Point 209°C to 213°C
Color Green to Yellow
Infrared Spectrum Authentic
Quantity 5 g
SMILES COC1=CC(=CNNC=C2C=C(OC)C(=O)C(OC)=C2)C=C(OC)C1=O
IUPAC Name 4-({2-[(3,5-dimethoxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]hydrazin-1-yl}methylidene)-2,6-dimethoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 360.37
Formula Weight 360.36
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Syringaldazine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.