missing translation for 'onlineSavingsMsg'
Learn More

Syringaldehyde, TRC

Product Code. 30415011
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30415011 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30415011

Brand: Toronto Research Chemicals TRCS920045100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Syringaldehyde
CAS 134-96-3
Molecular Formula C9 H10 O4
Quantity 100 g
Synonym Benzaldehyde','4-hydroxy-3,5-dimethoxy-, 4-Hydroxy-3,5-dimethoxybenzaldehyde, 2,6-Dimethoxy-4-formylphenol, 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Formyl-2,6-dimethoxyphenol, 4-Formylsyringol, Cedar aldehyde, Gallaldehyde 3,5-dimethyl ether, NSC 41153, SM 707, Syringaldehyde, Syringic aldehyde, VND 3207
InChI Formula InChI=1S/C9H10O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-5,11H,1-2H3
SMILES COc1cc(C=O)cc(OC)c1O
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.