missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syringaldehyde, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Syringaldehyde |
| CAS | 134-96-3 |
| Molecular Formula | C9 H10 O4 |
| Quantity | 1 g |
| Synonym | Benzaldehyde','4-hydroxy-3,5-dimethoxy-, 4-Hydroxy-3,5-dimethoxybenzaldehyde, 2,6-Dimethoxy-4-formylphenol, 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Formyl-2,6-dimethoxyphenol, 4-Formylsyringol, Cedar aldehyde, Gallaldehyde 3,5-dimethyl ether, NSC 41153, SM 707, Syringaldehyde, Syringic aldehyde, VND 3207 |
| InChI Formula | InChI=1S/C9H10O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-5,11H,1-2H3 |
| SMILES | COc1cc(C=O)cc(OC)c1O |
| IUPAC Name | 4-hydroxy-3,5-dimethoxybenzaldehyde |
| Molecular Weight (g/mol) | 182.17 |
| Formula Weight | 182.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction