missing translation for 'onlineSavingsMsg'
Learn More

Syringic Acid, TRC

Product Code. 30134719
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134719 50 g 50g
17924825 5 g 5g
17990615 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134719 Supplier Toronto Research Chemicals Supplier No. TRCS92002550G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-57-4
Molecular Formula C9 H10 O5
Molecular Weight (g/mol) 198.17
Synonym Benzoic acid, 4-hydroxy-3,5-dimethoxy-, Syringic acid (6CI), 2,6-Dimethoxy-4-carboxyphenol, 3,5-Dimethoxy-4-hydroxybenzoic acid, 4-Hydroxy-3,5-dimethoxybenzoic acid, Cedar acid, Gallic acid 3,5-dimethyl ether, NSC 2129
IUPAC Name 4-hydroxy-3,5-dimethoxybenzoic acid
SMILES COc1cc(cc(OC)c1O)C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H10O5/c1-13-6-3-5(9(11)12)4-7(14-2)8(6)10/h3-4,10H,1-2H3,(H,11,12)
Formula Weight 198.0528
Percent Purity >95%
Chemical Name or Material Syringic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.