missing translation for 'onlineSavingsMsg'
Learn More

Syringol Gentiobioside, TRC

Product Code. 30007119
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007119 0.5 mg 0.5mg
30136011 5 mg 5mg
30138312 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007119 Supplier Toronto Research Chemicals Supplier No. TRCG5969250.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1416253-72-9
Molecular Formula C20 H30 O13
Molecular Weight (g/mol) 478.444
Synonym (2S,3R,4S,5S,6R)-2-(2,6-Dimethoxyphenoxy)-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol, Syringol Gentobioside
IUPAC Name (2R,3R,4S,5S,6R)-2-[[(2R,3S,4S,5R,6S)-6-(2,6-dimethoxyphenoxy)-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES COc1cccc(OC)c1O[C@@H]2O[C@H](CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C20H30O13/c1-28-8-4-3-5-9(29-2)18(8)33-20-17(27)15(25)13(23)11(32-20)7-30-19-16(26)14(24)12(22)10(6-21)31-19/h3-5,10-17,19-27H,6-7H2,1-2H3/t10-,11-,12-,13-,14+,15+,16-,17-,19-,20+/m1/s1
Formula Weight 478.169
Percent Purity >95%
Chemical Name or Material 2,6-Dimethoxyphenyl-6-O- b-D-glucopyranosyl b-D-Glucopyranoside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.