Learn More
CELLPATH SYSTEM II+ BIOPSY CASSETTE - WHITE LASER (THREADED)
Product Code. 30226253
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 2000
Product Code. Color Unit Size
30226253 White Pack of 2000
1 options
Product Code. 30226253 Supplier CELLPATH Supplier No. EAM140272B
  • ONLINE EXCLUSIVE PRICE
    586.86 €
    248.22 € / Pack of 2000
    Save 338.64 €
    57% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System II+ Laser Biopsy Cassette comes with a separate white lid that is easily attached via the use of a hinge. Incorporating 0.9 x 0.9 mm apertures, the biopsy cassette offers improved specimen security for small biopsy and tissue fragments.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Product Type Cassette