Learn More
CELLPATH SYSTEM II CASSETTE LID - STAINLESS STEEL
Product Code. 30226269
Change view
Click to view available options
Color:
Gray
Unit Size:
Pack of 25
Product Code. Color Unit Size
30226269 Gray Pack of 25
1 options
Product Code. 30226269 Supplier CELLPATH Supplier No. EAX011408A
  • ONLINE EXCLUSIVE PRICE
    300.48 €
    119.32 € / Pack of 25
    Save 181.16 €
    60% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System II Stainless Steel Cassette Lid is compatible with the System II Cassette. Made of re-useable stainless-steel, a disposable plastic version is also available depending on your requirements.
The System II Cassette Lid is compatible with the System II Cassette.
Specifications
Color Gray
Product Type Cassette