Learn More
CELLPATH SYSTEM II HEX - CASSETTE
Product Code. 30226230
Change view
Click to view available options
Unit Size:
Pack of 1000
Pack of 2000
Material:
Laser Threaded
Loose
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Material
30226230 Pack of 2000 -
30226228 Pack of 1000 Loose
30226231 Pack of 2000 Laser Threaded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226230 Supplier CELLPATH Supplier No. EAD010272B
  • ONLINE EXCLUSIVE PRICE
    356.41 €
    150.75 € / Pack of 2000
    Save 205.66 €
    57% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The innovative honeycomb design of the The System II Hex Cassette maximises the open surface area compared to traditional round perforations, increasing reagent flow through for improved processing and a more rigid block when sectioning.
  • The System II Hex Cassette benefits from an optimised writing surface as well as extra thick side walls that increase stability and rigidity, essential for high-quality sectioning. Supplied loose, threaded (for easy loading of Leica and Sakura printers) or pre-loaded in hopper tubes (for easy loading of Thermo Scientific cassette printers).
  • Available in a range of colours, the System II Hex Cassette can be closed by using one of our plastic or stainless-steel lids.
  • Key Benefits:
  • Hex design maximises open surface area to increase the fluid exchange
  • Optimised writing surface for printing high-quality text, 1D and 2D barcodes
  • Writing surface suitable for marking with a CellMark marker pen and CellMark 2B pencil
  • Large colour range available
Specifications
Color White
No. of Compartments Threaded
Product Type Cassette