Learn More
CELLPATH SYSTEM II HEX - CASSETTE
Product Code. p-200113598
Click to view available options
Material:
Laser Threaded
Loose
Unit Size:
Pack of 1000
Pack of 2000
Product Code. 30226228
Brand: CELLPATH EAD010203A
Edge
The innovative honeycomb design of the The System II Hex Cassette maximises the open surface area compared to traditional round perforations, increasing reagent flow through for improved processing and a more rigid block when sectioning.
  • The System II Hex Cassette benefits from an optimised writing surface as well as extra thick side walls that increase stability and rigidity, essential for high-quality sectioning. Supplied loose, threaded (for easy loading of Leica and Sakura printers) or pre-loaded in hopper tubes (for easy loading of Thermo Scientific cassette printers).
  • Available in a range of colours, the System II Hex Cassette can be closed by using one of our plastic or stainless-steel lids.
  • Key Benefits:
  • Hex design maximises open surface area to increase the fluid exchange
  • Optimised writing surface for printing high-quality text, 1D and 2D barcodes
  • Writing surface suitable for marking with a CellMark marker pen and CellMark 2B pencil
  • Large colour range available
Specifications
Color White
Material Loose
Product Type Cassette