Learn More
CELLPATH SYSTEM II+ MICRO BIOPSY CASSETTE - WHITE LASER (THREADED)
Product Code. 30226254
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 2000
Product Code. Color Unit Size
30226254 White Pack of 2000
1 options
Product Code. 30226254 Supplier CELLPATH Supplier No. EAM150272B
  • ONLINE EXCLUSIVE PRICE
    586.86 €
    248.22 € / Pack of 2000
    Save 338.64 €
    57% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System II+ Micro Biopsy Laser Cassette incorporates an inner chamber with 0.38 x 0.38 mm apertures. The inner chamber has rounded corners that enable easy handling of small samples using forceps.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Product Type Cassette