Learn More
CELLPATH SYSTEM II+ ROUND CASSETTE
Product Code. 30226251
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226251 Threaded Pack of 2000
30226249 Loose Pack of 1000
30226250 Hoppered Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226251 Supplier CELLPATH Supplier No. EAM080272B
  • ONLINE EXCLUSIVE PRICE
    625.01 €
    248.22 € / Pack of 2000
    Save 376.79 €
    60% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System II+ Round Cassette incorporates a round 2.4 mm diameter holes facilitates increased reagent flow in a traditional cassette.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Threaded
Product Type Cassette