Learn More
CELLPATH SYSTEM II+ ROUND CASSETTE - WHITE LASER (THREADED)
Product Code. 30226255
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 2000
Product Code. Color Unit Size
30226255 White Pack of 2000
1 options
Product Code. 30226255 Supplier CELLPATH Supplier No. EAM160272B
  • ONLINE EXCLUSIVE PRICE
    625.01 €
    248.22 € / Pack of 2000
    Save 376.79 €
    60% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System II+ Round Laser Cassette incorporates round holes with a 2.4 mm diameter for increased reagent flow. Its separate white lid easily attaches using a hinge, and its robust locking mechanism closes with a positive 'click' and ensures the specimen remains secure during processing.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Product Type Cassette