Learn More
CELLPATH SYSTEM II+ SLOTTED CASSETTE
Product Code. 30226241
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226241 Hoppered Pack of 3000
30226240 Loose Pack of 1000
30226242 Threaded Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226241 Supplier CELLPATH Supplier No. EAM050205H
  • ONLINE EXCLUSIVE PRICE
    939.93 €
    373.29 € / Pack of 3000
    Save 566.64 €
    60% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Incorporating 1.1 mm wide slots, the System II+ Slotted Cassette increases reagent flow, facilitating improved consistency and quality of processed specimens. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Hoppered
Product Type Cassette