Learn More
CELLPATH SYSTEM III BIOPSY CASSETTE
Product Code. 30226237
Change view
Click to view available options
Material:
Loose
Threaded
Unit Size:
Pack of 2000
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226237 Threaded Pack of 2000
30226236 Loose Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30226237 Supplier CELLPATH Supplier No. EAI060272B
  • ONLINE EXCLUSIVE PRICE
    625.01 €
    248.22 € / Pack of 2000
    Save 376.79 €
    60% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System III Biopsy Cassette comes with an attached lid with an easy to break-off hinge. Incorporating square apertures of 0.9 mm by 0.9 mm, the Biopsy Cassette ensures safe accommodation of smaller biopsy specimens, whilst also enabling good levels of fluid transfer.
  • The locking mechanism of the cassette ensures maximum tissue security during processing.
  • The cassette has a precisely engineered writing surface which optimises the print quality on inkjet and thermal printers, including the Primera, Leica and Sakura Printers.
  • Key Benefits:
  • Compatible with Primera, Leica and Sakura Printers
  • Optimised writing surface
  • Increased reagent flow
  • Integral fixed frangible lid
  • High rigidity and stability
Specifications
Color White
Material Threaded
Product Type Cassette