Learn More
Velp Scientifica™ System Support
Product Code. 13445509 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
DK20 and DK42/26
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
13445509 DK20 and DK42/26 Each
1 options
Product Code. 13445509 Supplier Velp Scientifica™ Supplier No. A00000190
  • ONLINE EXCLUSIVE PRICE
    563.00 €
    541.50 € / Pack of 1
    Save 21.50 €
    3% Off
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Velp Scientifica™ System Support is designed for use with a variety of Kjeldahl Digestion Units.
  • Required accessory with DK Series Digestion Unit
Specifications
Product Type System Support
For Use With (Equipment) DK20 and DK42/26