missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Systemin
Specifications
Specifications
| CAS | 137181-56-7 |
| Content And Storage | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Molecular Weight (g/mol) | 2010.3 g/mol |
| Product Type | Natural Products |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?