missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SZL P1-41
GREENER_CHOICE

Product Code. 18795668
Change view
Click to view available options
:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18795668 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18795668 Supplier Tocris Bioscience™ Supplier No. 5076/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Skp2 inhibitor

SZL P1-41 is a Skp2 inhibitor; thought to prevent assembly of Skp2-Skp1 complexes. Selectively suppresses Skp2 SCF E3 ligase activity but exhibits no effect on the activity of other SCF complexes. Also inhibits Skp2-mediated p27 and Akt ubiquitination in vivo and in vitro. Suppresses the survival of cancer cells and cancer stem cells by triggering cell senescence and inhibiting glycolysis. Exhibits antitumor effects in multiple animal models and cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Benzothiazolyl)-6-ethyl-7-hydroxy-8-(1-piperidinylmethyl)-4H-1-benzopyran-4-one
CAS 222716-34-9
Quantity 10 mg
Molecular Formula C24H24N2O3S
Formula Weight 420.52
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.