Learn More
IKA™ T 18 mini digital Solution
Product Code. 31454121 Shop All IKA Products
Change view
Click to view available options
Plug Type:
CH
EU
UK
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type
31454121 CH
31454107 EU
31454110 UK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31454121 Supplier IKA™ Supplier No. 0010015140
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The T 18 mini digital Solution provides a complete starter solution to start your homogenisation journey, complete with a dispersing tool, shaft and stand for basic homogenisation requirements with a compact size and smart features.
T 18 mini digital is an economical high-performance dispersing instrument with a wide speed range of 3000 - 25,000 rpm for working volumes of up to 1.5 L (H2O). It enables the user to work at high circumferential speeds even with small rotor diameters. The compact design of this disperser allows for operation in the tightest of spaces. A large TFT display ensures easy readability from a greater distance, even from outside a fume hood. This setup is best suited for simple applications homogenizing samples with low viscosity. The T 18 mini digital Solution provides a complete starter solution to start your homogenisation journey, complete with a dispersing tool, shaft and stand for basic homogenisation requirements with a compact size and smart features.
Scope of delivery
  • T 18 mini digital ULTRA-TURRAX™
  • S 18 N - 19 G Dispersing tool
  • R 1826 Plate stand
  • R 182 Boss head clamp
TRUSTED_SUSTAINABILITY
Specifications
Product Type Homogenizer Package
Speed ≤460 rpm
Voltage 220/240 V
Packaging Type Individual Pack
Plug Type CH