Learn More
IKA™ T 25 easy clean control Solution
Product Code. 31454114 Shop All IKA Products
Change view
Click to view available options
Plug Type:
CH
EU
UK
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
31454114 EU Each
31454096 UK -
31454098 CH -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31454114 Supplier IKA™ Supplier No. 0010015172
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 08-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The T 25 easy clean control Solution features tools designed for quick, residue-free cleaning and supports smart automation with data logging. Ideal for precise, hygienic homogenization in industries such as pharmaceuticals, cosmetics, chemicals, and food production.
The T 25 easy clean control is a high-performance dispersing instrument with electronic speed control from 3,000 - 25,000 rpm for working volumes of 1 - 2,000 mL (H2O). The new tool S 25 EC-T-C-25G allows temperature measurement and operation time detection through RFID. Sample overheating is prevented by continuous temperature monitoring and the setting of temperature limits. The detection of operation time serves to guarantee regular maintenance.
  • Continuous temperature measurement with patented temperature sensing tool including an integrated temperature sensor
  • Operation time detection through RFID
  • Quick and easy cleaning of the easy clean dispersing tools, no disassembling required
  • Brushless motor guarantees dust-free and silent operation
  • USB interface for control and documentation via labworldsoft™
  • Timer/counter function
  • Increased safety through tool recognition and IP-class
  • Remote control with WiCo (not incl. with delivery)
All S 25 tools are compatible with T 25 easy clean control. The T 25 easy clean control Solution provides a complete solution to start your homogenisation journey, complete with a dispersing tool, shaft and stand, ideal for applications in industries working in ambient conditions that require a high degree of hygience and compliance. This solution is also extremely versatile and suitable for use for a wide range of industries such as chemicals, cosmetics, pharmaceuticals, agriculture and more, especially in applications involving heat-sensitive products and requiring automation, device-to-device communication and data logging.
Scope of delivery
  • T 25 easy clean control ULTRA-TURRAX™
  • S 25 EC - T - C - 25 G Dispersing tool
  • R 1826 Plate stand
  • R 182 Boss head clamp
TRUSTED_SUSTAINABILITY
Specifications
Product Type Homogenizer Package
Speed ≤460 rpm
Voltage 220/240 V
Packaging Type Individual Pack
Plug Type EU