missing translation for 'onlineSavingsMsg'
Learn More

t-Butyl4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate, ≥95%, Thermo Scientific™

Product Code. 10601275
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10601275 250 mg Amber Glass Bottle 250mg
10527853 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10601275 Supplier Thermo Scientific Maybridge Supplier No. CC31023CB
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 365413-31-6
Molecular Formula C16H24N2O4S
Molecular Weight (g/mol) 340.438
MDL Number MFCD06659069
InChI Key LBRUPXMVELEXIV-UHFFFAOYSA-N
Synonym tert-butyl 4-4-ethoxycarbonyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, ethyl 2-1-tert-butoxycarbonyl piperidin-4-yl thiazole-4-carboxylate, tert-butyl 4-4-ethoxycarbonyl-1,3-thiazol-2-yl piperidine-1-carboxylate, 1-boc-4-4-ethoxycarbonyl-thiazol-2-yl-piperidine, tert-butyl 4-4-ethoxycarbonyl-1,3-thiazol-2-yl, tert-butyl 4-4-ethoxycarbonylthiazol-2-yl piperidine-1-carboxylate, 1,1-dimethylethyl 4-4-ethoxycarbonyl-2-thiazolyl-1-piperidine-carboxylate, 1,1-dimethylethyl 4-4-ethoxycarbonyl-2-thiazolyl-1-piperidinecarboxylate, 4-4-ethoxycarbonyl-thiazol-2-yl-piperidine-1-carboxylic acid t-butyl ester, 4-4-ethoxycarbonyl-thiazol-2-yl-piperidine-carboxylic acid tert-butyl ester
PubChem CID 2795532
IUPAC Name ethyl 2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazole-4-carboxylate
SMILES CCOC(=O)C1=CSC(=N1)C2CCN(CC2)C(=O)OC(C)(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 340.44
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.