Learn More
Thermo Scientific™ T-REx™ Tango™ P2RY14-bla U2OS Cells
Description
T-REx™ Tango™ P2RY14-bla U2OS cells contain the human purinergic receptor P2Y14 (P2RY14) under doxycycline inducible control linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The T-REx™ Tango™ P2RY14-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of UDP-glucose. In addition, T-REx™ Tango™ P2RY14-bla U2OS cells have been tested for assay performance under variable conditions.
Specifications
Specifications
| Accession Number | NM_014879.2 |
| Assay Entry | Cell-based beta-arrestin recruitment |
| Cell State | Dividing Cells |
| Content And Storage | Tango™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Cytoplasmic Tail | Native Tail |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Human Protein Class | G-Protein Coupled Receptor (GPCR) |
| Population Status | Single Cell Clone Origin |
| Product Line | Tango |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.