Learn More
Thermo Scientific™ T-valve Assembly for LN2 Supply Tank
Product Code. 15152889 Shop All Thermo Scientific Products
Change view
Click to view available options
Manual valve and T fitting
Product Code.
15152889 Manual valve and T fitting
1 options
Product Code. 15152889 Supplier Thermo Scientific™ Supplier No. 4000290
  • ONLINE EXCLUSIVE PRICE
    389.00 €
    375.00 € / Pack of 1
    Save 14.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Connect two hoses to a LN2 tank with this T-valve assembly, which includes a manual valve and T-fitting.
Connect two hoses to a LN2 tank with this Thermo Scientific™ T-valve assembly for LN2 supply tank, which includes a manual valve and T-fitting.
TRUSTED_SUSTAINABILITY
Specifications
Description Manual valve and T fitting
Type Manual Valve and T-Fitting
For Use With (Equipment) CryoPlus 1,2,3,4
Defrost Manual