missing translation for 'onlineSavingsMsg'
Learn More

T16Ainh - A01, Tocris Bioscience™

Product Code. 18003842
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003842 10 mg 10mg
18025212 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18003842 Supplier Tocris Bioscience Supplier No. 4538/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ca2+-activated Cl- channel transmembrane protein 16A (TMEM16A) inhibitor

Inhibitor of Ca2+-dependent Cl- channel (CaCC) transmembrane protein 16A (TMEM16A) (IC50 = 1.8 μM in A253 salivary gland epithelial cells). Inhibits EGF-induced increases in CaCC currents; blocks proliferation of tumor cells in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552309-42-9
Molecular Formula C19H20N4O3S2
Molecular Weight (g/mol) 416.514
InChI Key QSIYTNYMBWYHAA-UHFFFAOYSA-N
Synonym t16a inh-a01, t16ainh-a01, 2-5-ethyl-4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl thio-n-4-4-methoxyphenyl thiazol-2-yl acetamide, 2-5-ethyl-1,6-dihydro-4-methyl-6-oxo-2-pyrimidinyl thio-n-4-4-methoxyphenyl-2-thiazolyl acetamide, 2-5-ethyl-4-hydroxy-6-methylpyrimidin-2-yl sulfanyl-n-4-4-methoxyphenyl-1,3-thiazol-2-yl acetamide, t16ainh-a hplc, 2-5-ethyl-4-hydroxy-6-methylpyrimidin-2-ylthio-n-4-4-methoxyphenyl thiazol-2-yl acetamide, 2-5-ethyl-4-methyl-6-oxo-3h-pyrimidin-2-yl sulfanyl-n-4-4-methoxyphenyl-1,3-thiazol-2-yl acetamide, 2-5-ethyl-6-methyl-4-oxo-1h-pyrimidin-2-yl sulfanyl-n-4-4-methoxyphenyl-1,3-thiazol-2-yl acetamide, n-4-4-methoxyphenyl thiazole-2-yl-2-4-hydroxy-5-ethyl-6-methylpyrimidine-2-ylthio acetamide
PubChem CID 3193184
IUPAC Name 2-[(5-ethyl-6-methyl-4-oxo-1H-pyrimidin-2-yl)sulfanyl]-N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]acetamide
SMILES CCC1=C(NC(=NC1=O)SCC(=O)NC2=NC(=CS2)C3=CC=C(C=C3)OC)C

Specifications

Quantity 10 mg
Formula Weight 416.52
Percent Purity >98%
Chemical Name or Material T16Ainh - A01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.