missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ T3

Product Code. 18726236
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18726236 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18726236 Supplier R&D Systems™ Supplier No. 6666/50
  • ONLINE EXCLUSIVE PRICE
    87.00 €
    81.90 € / 50mg
    Save 5.10 €
    5% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Thyroid hormone; also promotes differentiation of oligodendroglial precursor cells

T3 is a thyroid hormone. Promotes differentiation of oligodendroglial precursor cells in neonatal rats and promotes maturation of hiPSC-derived cardiomyocytes. Also specifies cone cell subtype in retinal cell differentiation protocols. T3 is used in protocols to generate cortical spheroids from hPSCs.

For more information about how T3 may be used, see our protocol:

Generation of β cells from hPSCs
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-(4-Hydroxy-3-iodophenyl-3,5-diiodo-L-tyrosine
CAS 6893-02-3
Quantity 50 mg
Product Line Tocris
Target Compounds for Stem Cell Differentiation
Molecular Formula C15H12I3NO4
Formula Weight 650.97
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.