missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ T3 CLK

Description
T3 CLK is a potent and selective pan CDC-like kinase (CLK) inhibitor (IC50 values are 0.67 nM, 15 nM, and 110 nM for CLK1, CLK2, and CLK3 protein kinases, respectively). T3 CLK also inhibits DYRK1A/B (IC50 values are 230 nM and 260 nM for DYRK1B and DYRK1A, respectively in cellular assays). Exhibits dose-dependent alternative splicing effects in HCT116 colorectal cancer cells. T3 CLK is cell permeable, stable and non-toxic.
Specifications
Specifications
| Chemical Name or Material | 4-[1,1-Dimethyl-2-(4-methyl-1-piperazinyl)-2-oxoethyl]-N-[6-(4-pyridinyl)imidazo[1,2-a]pyridin-2-yl]benzamide |
| CAS | 2109805-56-1 |
| Quantity | 10 mg |
| Target | Cdc2-like Kinase Inhibitors |
| Molecular Formula | C28H30N6O2 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?