missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T4 DNA Ligase, LC (1 U/μL)

Product Code. 10786591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 500 units
Unit Size:
2 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10786591 2 x 500 units 2 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10786591 Supplier Thermo Scientific™ Supplier No. EL0016
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.

Catalyze the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA with the Thermo Scientific™ T4 DNA Ligase. The enzyme repairs single-strand nicks in duplex DNA, RNA, or DNA/RNA hybrids. It also joins DNA fragments with either cohesive or blunt termini, but has no activity on single-stranded nucleic acids.

The T4 DNA Ligase requires ATP as a cofactor.

Highlights

  • Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
  • Fast – sticky-end ligation is completed in 10 minutes at room temperature
  • Supplied with PEG solution for efficient blunt-end ligation

Applications

  • Cloning of restriction enzyme generated DNA fragments
  • Cloning of PCR products
  • Joining of double-stranded oligonucleotide linkers or adaptors to DNA
  • Site-directed mutagenesis
  • Amplified fragment length polymorphism (AFLP)
  • Ligase-mediated RNA detection (see Reference 3)
  • Nick repair in duplex DNA, RNA or DNA/RNA hybrids
  • Self-circularization of linear DNA.

Notes

  • Binding of T4 DNA Ligase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 70°C for 5 min or 65°C for 10 minutes and chill on ice prior to electrophoresis.
  • The volume of the ligation reaction mixture should not exceed 10% of the competent cell volume in the transformation process.
  • Prior to electro-transformation, remove T4 DNA Ligase from the ligation mixture by spin column or chloroform extraction. The extracted DNA can be further precipitated with ethanol.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 U/μL
Enzyme DNA Ligase
Compatible Buffer 10X T4 DNA Ligase Buffer
Quantity 2 x 500 units
Product Type T4 DNA Ligase
compliance-icons
Product Identifier
  • T4 DNA Ligase, LC (1 U/µL)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: (R*,R*)-1,4-dimercapto-2,3-butanediol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.