missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Table Paper Boards

Product Code. 15972810 Shop All Sartorius Products
Change view
Click to view available options
Dimensions (L x W):
210 x 297 mm (8.3 x 11.7 in.)
580 x 580 mm (22.8 x 22.8 in.)
Unit Size:
Pack of 100
Pack of 200
Product Code. Dimensions (L x W) unitSize
15972810 210 x 297 mm (8.3 x 11.7 in.) Pack of 200
15982810 580 x 580 mm (22.8 x 22.8 in.) Pack of 100
2 options
This item is not returnable. View return policy

Product Code. 15972810

Brand: Sartorius FT2H182210297

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Table Paper Boards for the filtration of cooking and transformer oils and as base paper for further impregnation with certain reagents. The grades with a wet bursting strength 30 kPa are identified as wet strengthened and suitable for pressure or vacuum filtration.

Filter Boards
GradeBasis weight g/m2Thickness in mm
0.904201.30
12204751.10
13393150.63
1514600.90
1523800.90
152 A3401.00
15222800.60
1577001.80
16002500.50
16321500.63
167 3603600.90
5114901.25
641000.22
A 1401400.25
A 2502500.40
C 1601600.30
C 2502500.43
C 2512500.55
C 3003000.52
C 3503500.63
C 350 L3600.75
C 4504500.95
K 125201.50
LF 13600.90
M 2702700.62
M 6006001.60
S 1651650.33
S 5005001.00
SB 21550.34
SEK 5505501.25
SEK 7707701.00
TRUSTED_SUSTAINABILITY

Specifications

Format Sheet
Grade H182
Type Filter Board
Dimensions (L x W) 210 x 297 mm (8.3 x 11.7 in.)
Fit for Purpose Certified Quality
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.