missing translation for 'onlineSavingsMsg'
Learn More

Tacrine hydrochloride (hydrate), MedChemExpress

Product Code. 16488876
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16488876

Brand: Medchem Express HYB2244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tacrine hydrochloride (hydrate) is an inhibitor of both acetyl (AChE) and butyryl-cholinestrase (BChE) with IC50s of 31 nM and 25.6 nM, respectively.

Tacrine hydrochloride (hydrate) is an inhibitor of both acetyl (AChE) and butyryl-cholinestrase (BChE) with IC50s of 31 nM and 25.6 nM, respectively.

Specifications

Chemical Name or Material Tacrine hydrochloride (hydrate)
CAS 206658-92-6
Color White
Purity Grade Notes Research
Molecular Formula C13H14N2.xHCl.xH2O
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (Need ultrasonic) ∣DMSO : 32 mg/mL (Need ultrasonic and warming)
SMILES NC1=C(CCCC2)C2=NC3=CC=CC=C31.[H]Cl.[H]O[H].[x].[x]
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.