missing translation for 'onlineSavingsMsg'
Learn More

Tacrolimus, MedChemExpress

Product Code. 16495838
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495838 10 mg 10mg
16416316 100 mg 100mg
16411106 200 mg 200mg
16481987 50 mg 50mg
16411706 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495838 Supplier Medchem Express Supplier No. HY13756
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tacrolimus (FK506), a macrocyclic lactone, binds to FK506 binding protein (FKBP) to form a complex. Tacrolimus inhibits calcineurin phosphatase, which inhibits T-lymphocyte signal transduction and IL-2 transcription. Immunosuppressive properties.

Tacrolimus (FK506), a macrocyclic lactone, binds to FK506 binding protein (FKBP) to form a complex. Tacrolimus inhibits calcineurin phosphatase, which inhibits T-lymphocyte signal transduction and IL-2 transcription. Immunosuppressive properties.

Specifications

Chemical Name or Material Tacrolimus
CAS 104987-11-3
Color White
Purity Grade Notes Research
Molecular Formula C44H69NO12
Quantity 10 mg
Synonym FK506 Fujimycin FR900506
Solubility Information DMSO : ≥ 150 mg/mL (186.56 mM)
SMILES O=C([C@@](CCCC1)([H])N1C(C([C@@]2(O)[C@H](C)C[C@H](OC)[C@@](O2)([H])[C@@H](OC)C[C@@H](C)C/C(C)=C/[C@H]3CC=C)=O)=O)O[C@H](/C(C)=C/[C@H]4C[C@@H](OC)[C@H](O)CC4)[C@H](C)[C@@H](O)CC3=O
Molecular Weight (g/mol) 804.02
Formula Weight 804.02
Percent Purity 95.2%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.