missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ion Torrent™ Tag Sequencing Barcode Set 25-48
Description
- The Tag Sequencing Barcode Set 25-48 provides a set of 24 unique barcode adapters specifically designed for optimal performance in Oncomine™ cfDNA assays. The barcode adapters are compatible with the Ion S5™, Ion PGM™, and Ion Proton™ sequencing systems. When used in combination with kits utilizing tag sequencing technology, this barcode set enables users to pool up to 24 amplicon libraries and to conduct multiplexed next-generation sequencing (NGS) analysis, reducing the sequencing cost per sample.
- Enables multiplexing of numerous amplicon library samples on a single sequencing chip with the use of robust molecular barcodes
- Both sequence-optimized and flow-optimized for equal representation of all barcodes in a pool and more economical multiplexed sequencing runs
- Compatible with library construction kits utilizing tag sequencing technology
- Multiplexing with barcoded libraries leads to more cost-effective runs by permitting up to 24 samples per run (or more if combined with Tag Sequencing BC Set 1-24, Cat. No. A31830), significantly decreasing the cost and handling requirements of sequencing. Each barcode was individually tested to minimize representation bias.
- Barcode adapters contain sequences that are optimized to provide equal representation of all barcodes in a pool; they require a minimal number of flows to interrogate resulting in more economical multiplexed sequencing runs
Specifications
Specifications
| Product Type | Oligonucleotides, Primers |
| Content And Storage | 10 μL each Tag Sequencing Barcodes 24 thru 48, 10 reactions/barcode Store in freezer (-5 to -30°C). |
| Shipping Condition | Dry Ice |
| Primer | Custom Primer |
| Quantity | 1 Set |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?