missing translation for 'onlineSavingsMsg'
Learn More

TAK-285, MedChemExpress

Product Code. 16496358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496358 10 mg 10mg
16488947 5 mg 5mg
16432477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496358 Supplier Medchem Express Supplier No. HY15196
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TAK-285 is a potent, selective, ATP-competitive and orally active HER2 and EGFR(HER1) inhibitor with IC50 of 17 nM and 23 nM, respectively. TAK-285 is >10-fold selectivity for HER1/2 than HER4, and less potent to MEK1/5, c-Met, Aurora B, Lck, CSK etc. TAK-285 has effective antitumor activity. TAK-285 can cross the blood-brain barrier (BBB).

TAK-285 is a potent, selective, ATP-competitive and orally active HER2 and EGFR(HER1) inhibitor with IC50 of 17 nM and 23 nM, respectively. TAK-285 is >10-fold selectivity for HER1/2 than HER4, and less potent to MEK1/5, c-Met, Aurora B, Lck, CSK etc. TAK-285 has effective antitumor activity. TAK-285 can cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material TAK-285
CAS 871026-44-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H25ClF3N5O3
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (91.25 mM)
SMILES ClC(C=C(NC1=NC=NC2=C1N(CCNC(CC(C)(C)O)=O)C=C2)C=C3)=C3OC4=CC=CC(C(F)(F)F)=C4
Molecular Weight (g/mol) 547.96
Formula Weight 547.96
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.