missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK-715, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
TAK-715 is an orally active and potent p38 MAPK inhibitor with IC50s of 7.1 nM, 200 nM for p38α and p38β, respectively. TAK-715 inhibits casein kinase I (CK1δ/ε) to regulate activation of Wnt/β-catenin signaling. TAK-715 shows good significant efficacy in a rat arthritis model.
Specifications
Specifications
| Chemical Name or Material | TAK-715 |
| CAS | 303162-79-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H21N3OS |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (250.31 mM) |
| SMILES | O=C(C1=CC=CC=C1)NC2=NC=CC(C3=C(N=C(S3)CC)C4=CC=CC(C)=C4)=C2 |
| Molecular Weight (g/mol) | 399.51 |
| Formula Weight | 399.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction