missing translation for 'onlineSavingsMsg'
Learn More

Talarozole, MedChemExpress

Product Code. 16496476
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496476 100 mg 100mg
16470216 2 mg 2mg
16448687 5 mg 5mg
16402227 50 mg 50mg
16416088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496476 Supplier Medchem Express Supplier No. HY14531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Talarozole (R115866) is an oral systemic all-trans retinoic acid metabolism blocking agent (RAMBA) which increases intracellular levels of endogenous all-trans retinoic acid (RA). Talarozole inhibits both CYP26A1 and CYP26B1 with IC50s of 5.4 and 0.46 nM, respectively.

Talarozole (R115866) is an oral systemic all-trans retinoic acid metabolism blocking agent (RAMBA) which increases intracellular levels of endogenous all-trans retinoic acid (RA). Talarozole inhibits both CYP26A1 and CYP26B1 with IC50s of 5.4 and 0.46 nM, respectively.

Specifications

Chemical Name or Material Talarozole
CAS 201410-53-9
Color White
Purity Grade Notes Research
Molecular Formula C21H23N5S
Quantity 100 mg
Synonym R115866
Solubility Information DMSO : ≥ 36 mg/mL (95.36 mM)
SMILES CCC(CC)C(C1=CC=C(NC2=NC3=CC=CC=C3S2)C=C1)N4N=CN=C4
Molecular Weight (g/mol) 377.51
Formula Weight 377.51
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.