Learn More
Köttermann Tall Safety Storage Cabinet with Doors
Product Code. 17382129
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Door Style:
Folding, Manual-close
Hinged, Auto-close
No. of Shelves:
3
4
6
None
Includes:
1 Drip Tray
4 Internal Drawers
5 Internal Drawers
missing translation for 'unitSize':
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style No. of Shelves Includes missing translation for 'unitSize'
17382129 Hinged, Auto-close None 5 Internal Drawers Each
17362129 Hinged, Auto-close 3 1 Drip Tray Each
17372129 Hinged, Auto-close 6 1 Drip Tray Each
17312139 Hinged, Auto-close 4 1 Drip Tray Each
17302139 Folding, Manual-close None 4 Internal Drawers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Product Code. 17382129 missing translation for 'mfr' Köttermann missing translation for 'supplierNo' 37100031
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
  • Different versions that meet your needs
  • Additional storage space
  • Top safety for flammable hazardous substances (Type 90)
Specifications
Dimensions (D x W x H) 615 x 1200 x 1955 mm
Door Style Hinged, Auto-close
Includes 5 Internal Drawers
No. of Doors 2
No. of Shelves None