Learn More
Köttermann Tall Safety Storage Cabinet with Drawers
Product Code. 17322139
Change view
Click to view available options
No. of Doors:
1
2
No. of Shelves:
4
5
8
9
10
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Doors No. of Shelves Unit Size
17322139 1 4 Each
17332139 1 5 Each
17342139 2 8 Each
17362139 2 9 Each
17352139 2 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17322139 Supplier Köttermann Supplier No. 37100036
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
  • Different versions that meet your needs
  • Additional storage space
  • Top safety for flammable hazardous substances (Type 90)
Specifications
Dimensions (D x W x H) 853 x 450 x 1966 mm
Door Style Drawer
Includes 1 Drip Tray
No. of Doors 1
No. of Shelves 4