Learn More
Köttermann Tall Safety Storage Cabinet with Drawers
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
Brand: Köttermann 371-00038
Description
- Different versions that meet your needs
- Additional storage space
- Top safety for flammable hazardous substances (Type 90)
Specifications
1 Drip Tray | |
Drawer | |
2 |
853 x 815 x 1966 mm | |
8 |