missing translation for 'onlineSavingsMsg'
Learn More

Talsaclidine, TRC

Product Code. 30419799
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419799 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419799 Supplier Toronto Research Chemicals Supplier No. TRCT0056005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 147025-53-4
Molecular Formula C10 H15 N O
Quantity 5 mg
Synonym 1-Azabicyclo[2.2.2]octane, 3-(2-propyn-1-yloxy)-, (3R)-, 1-Azabicyclo[2.2.2]octane, 3-(2-propynyloxy)-, (R)- (9CI), (3R)-3-(2-Propyn-1-yloxy)-1-azabicyclo[2.2.2]octane, Talsaclidine
InChI Formula InChI=1S/C10H15NO/c1-2-7-12-10-8-9-3-5-11(10)6-4-9/h9-10H,3-6,8H2,1H3/t10-/m1/s1
SMILES CC#CO[C@@H]1CC2CCN1CC2
IUPAC Name (2R)-2-prop-1-ynoxy-1-azabicyclo[2.2.2]octane
Molecular Weight (g/mol) 165.23
Formula Weight 165.1154
Chemical Name or Material Talsaclidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.