missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Tamoxifen

Product Code. 18772729
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772729 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772729 Supplier R&D Systems™ Supplier No. 6342/100
  • ONLINE EXCLUSIVE PRICE
    143.23 €
    128.10 € / 100mg
    Save 15.13 €
    10% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Estrogen receptor partial agonist/antagonist

Tamoxifen is a selective estrogen receptor modulator (SERM). Exhibits antiestrogenic effects in mammary tissue and estrogen-like effects in bone. Anticancer. Tamoxifen citrate (Cat.No. 0999) is also available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-[(1Z)-1,2-Diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine
CAS 10540-29-1
Quantity 100 mg
Product Line Tocris
Target Estrogen and Related Receptor Modulators
Molecular Formula C26H29NO
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.