missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Tamoxifen citrate

Product Code. 18772774
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772774 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772774 Supplier R&D Systems™ Supplier No. 0999/100
  • ONLINE EXCLUSIVE PRICE
    128.91 €
    115.50 € / 100mg
    Save 13.41 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Estrogen receptor partial agonist/antagonist

Tamoxifen citrate is an estrogen receptor antagonist/partial agonist. Selective and potent inhibitor of mammalian sterol isomerase. Neuroprotective in female rats in vivo. Also high affinity agonist at the membrane estrogen receptor GPER. Tamoxifen (Cat. No. 6432) is also available.
TRUSTED_SUSTAINABILITY

Specifications

CAS 54965-24-1
Quantity 100 mg
Product Line Tocris
Molecular Formula C32H37NO8
Formula Weight 563.65
Purity >99%
Molecular Weight (g/mol) 563.65
Chemical Name or Material Tamoxifen citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.