missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen Citrate, TRC

Product Code. 30426604
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426604 500 mg 500mg
30413921 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426604 Supplier Toronto Research Chemicals Supplier No. TRCT006090500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tamoxifen citrate
CAS 54965-24-1
Molecular Formula C26 H29 N O . C6 H8 O7
Quantity 500 mg
Synonym Tamoxifen Citrate, Ethanamine','2-[4-[(1Z)-1,2-diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethyl-','2-hydroxy-1,2,3-propanetricarboxylate (1:1), Ethanamine','2-[4-(1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethyl-','(Z)-','2-hydroxy-1,2,3-propanetricarboxylate (1:1), Ethanamine','2-[4-[(1Z)-1,2-diphenyl-1-butenyl]phenoxy]-N,N-dimethyl-','2-hydroxy-1,2,3-propanetricarboxylate (1:1) (9CI), I.C.I.46474 citrate, ICI 46474, Istubal, Kessar, Noltam, Nolvadex, Nourytam, Soltamox, TMX, Tamofen, Tamoplex, Tamox-Puren, Tomaxasta, Valodex, Z-Tamoxifen citrate, Zemide
InChI Formula InChI=1S/C26H29NO.C6H8O7/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,4,19-20H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;
SMILES CC\C(=C(/c1ccccc1)\c2ccc(OCCN(C)C)cc2)\c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC Name 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 563.64
Formula Weight 563.2519
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.