Learn More
Tamoxifen, Tocris Bioscience™
Estrogen receptor partial agonist/antagonist
Brand:  Tocris Bioscience 6342/100
Encompass
Product Code. 15776329
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Tamoxifen is a selective estrogen receptor modulator (SERM). Exhibits antiestrogenic effects in mammary tissue and estrogen-like effects in bone. Anticancer. Tamoxifen citrate (Cat.No. 0999) is also available.
Specifications
Specifications
2-[4-[(1Z)-1,2-Diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine
10540-29-1
>99%