missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, TRC

Product Code. 30040971
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
10g
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040971 10 g 10g
18032230 2.5 g 2.5g
18091230 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040971 Supplier Toronto Research Chemicals Supplier No. TRCT00600010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10540-29-1
Formula Weight 371.2249
InChI Formula InChI=1S/C26H29NO/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3/h5-18H,4,19-20H2,1-3H3/b26-25-
IUPAC Name 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
Percent Purity >95%
SMILES CC\C(=C(/c1ccccc1)\c2ccc(OCCN(C)C)cc2)\c3ccccc3
Synonym (Z)-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine, ICI 47699, Mammaton, Novaldex, Tamoxifen, Z-Tamoxifen, trans-Tamoxifen;
Chemical Name or Material Tamoxifen
Grade HPLC
Show More Show Less

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.