Learn More
DWK Life Sciences Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 32
GL 45
GL 54
GL 60
Unit Size:
Pack of 10
Description
Tamper evident, blue polypropylene, linerless screw cap for wide mouth/short form DIN thread soda lime glass bottles.
- Tamper evident ring
- LInerless
- Short form DIN thread
Specifications
Specifications
| For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
| Material | Polypropylene |
| Quantity | 10/Pk. |
| Thread Size (Metric) | GL 60 |
| Diameter (Metric) | 78 mm |
| Autoclavable | Autoclavable |